Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290476.1 | NP_001277405.1 | 4 - 5 | 739 | 78 | |
NM_026167.4 | NP_080443.1 | 6 - 7 | 758 | 78 | ||
GenBank mRNA | AK004677.1 | - | 6 - 7 | 730 | 78 | |
AK122491.1 | - | 4 - 5 | 739 | 78 | ||
BC100393.1 | - | 5 - 6 | 723 | 30737426 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |