Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Narfl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026238.4 | NP_080514.3 | 9 - 10 | 1084 | 65 | |
GenBank mRNA | AK003464.1 | - | 4 - 5 | 417 | 65 | |
AK014038.1 | - | 9 - 10 | 1068 | 65 | ||
AK018548.1 | - | 9 - 10 | 1055 | 65 | ||
AK043694.1 | - | 9 - 10 | 1061 | 65 | ||
AK145408.1 | - | 9 - 10 | 1135 | 65 | ||
BC020468.1 | - | 9 - 10 | 1044 | 65 | ||
BC052830.1 | - | 9 - 10 | 1061 | 30040958 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |