Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030711.4 | NP_109636.1 | 7 - 8 | 1236 | 93 | |
GenBank mRNA | AB047552.1 | - | 6 - 7 | 1224 | 93 | |
AF227511.1 | - | 6 - 7 | 1212 | 93 | ||
AK029841.1 | - | 7 - 8 | 1235 | 93 | ||
AK030329.1 | - | 7 - 8 | 1237 | 93 | ||
AK036559.1 | - | 1237 | 93 | |||
AK040350.1 | - | 4 - 5 | 454 | 93 | ||
AK081181.1 | - | 7 - 8 | 1263 | 93 | ||
AK159406.1 | - | 7 - 8 | 1237 | 93 | ||
AK163954.1 | - | 7 - 8 | 1256 | 93 | ||
BC046610.1 | - | 7 - 8 | 1242 | 6397585 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |