Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282054.1 | NP_001268983.1 | 18 - 19 | 2475 | 102 | |
NM_031870.3 | NP_114076.1 | 17 - 18 | 2566 | 102 | ||
GenBank mRNA | AF178957.1 | - | 17 - 18 | 2270 | 102 | |
AF298655.1 | - | 17 - 18 | 2566 | 102 | ||
AK029837.1 | - | 18 - 19 | 2475 | 102 | ||
AK161474.1 | - | 17 - 18 | 2316 | 102 | ||
AY351585.1 | - | 17 - 18 | 2520 | 102 | ||
AY351587.1 | - | 6 - 7 | 1517 | 102 | ||
AY351590.1 | - | 9 - 10 | 1982 | 102 | ||
AY351591.1 | - | 10 - 11 | 2006 | 102 | ||
BC138163.1 | - | 17 - 18 | 2457 | 8861182 | 102 | |
BC145838.1 | - | 17 - 18 | 2457 | 40131141 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |