Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011139.2 | NP_035269.2 | 9 - 10 | 961 | 69 | |
GenBank mRNA | AK153697.1 | - | 1051 | 69 | ||
L14677.1 | - | 9 - 10 | 959 | 69 | ||
X51960.1 | - | 2 - 3 | 290 | 69 | ||
Z18537.1 | - | 9 - 10 | 962 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |