Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164696.1 | NP_001158168.1 | 2 - 3 | 209 | 67 | |
NM_001164697.1 | NP_001158169.1 | 2 - 3 | 205 | 67 | ||
NM_001164698.1 | NP_001158170.1 | 2 - 3 | 106 | 67 | ||
NM_011177.2 | NP_035307.2 | 1 - 2 | 217 | 67 | ||
GenBank mRNA | AB015206.1 | - | 2 - 3 | 193 | 67 | |
AK138218.1 | - | 2 - 3 | 206 | 67 | ||
BC031119.1 | - | 1 - 2 | 54 | 4954976 | 67 | |
EL956218.1 | - | 2 - 3 | 81 | 67 | ||
Y18723.1 | - | 1 - 2 | 217 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |