Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013610.2 | NP_038638.1 | 2 - 3 | 318 | 73 | |
GenBank mRNA | AK077602.1 | - | 2 - 3 | 337 | 73 | |
AK155741.1 | - | 2 - 3 | 328 | 73 | ||
AK159111.1 | - | 2 - 3 | 338 | 73 | ||
AK160045.1 | - | 2 - 3 | 325 | 73 | ||
AK171973.1 | - | 2 - 3 | 337 | 73 | ||
BC049600.1 | - | 2 - 3 | 305 | 73 | ||
BC118937.1 | - | 2 - 3 | 318 | 40091105 | 73 | |
U91513.1 | - | 2 - 3 | 318 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |