Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110809.2 | NP_001104279.1 | 1 - 2 | 67 | 75 | |
GenBank mRNA | AK011617.1 | - | 1 - 2 | 66 | 75 | |
AK018306.1 | - | 1 - 2 | 41 | 75 | ||
AK078828.1 | - | 1 - 2 | 65 | 75 | ||
AK169111.1 | - | 1 - 2 | 56 | 75 | ||
BC002265.1 | - | 1 - 2 | 38 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |