Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025888.5 | NP_080164.2 | 4 - 5 | 680 | 59 | |
GenBank mRNA | AK010404.1 | - | 3 - 4 | 443 | 59 | |
AK030202.1 | - | 4 - 5 | 682 | 59 | ||
AK088807.1 | - | 3 - 4 | 448 | 59 | ||
AK168440.1 | - | 4 - 5 | 671 | 59 | ||
BC036318.1 | - | 2 - 3 | 325 | 59 | ||
BC048161.1 | - | 3 - 4 | 464 | 59 | ||
BC049250.1 | - | 3 - 4 | 557 | 5042309 | 59 | |
BC052712.1 | - | 3 - 4 | 466 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |