Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026015.2 | NP_080291.1 | 2 - 3 | 197 | 67 | |
GenBank mRNA | AK003361.1 | - | 2 - 3 | 190 | 67 | |
AK007565.1 | - | 2 - 3 | 184 | 67 | ||
AK012098.1 | - | 2 - 3 | 198 | 67 | ||
AK013415.1 | - | 1 - 2 | 170 | 67 | ||
BC031796.1 | - | 2 - 3 | 176 | 4205598 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |