Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007601.3 | NP_031627.2 | 15 - 16 | 2085 | 70 | |
GenBank mRNA | BC139790.1 | - | 15 - 16 | 1805 | 40047690 | 70 |
X92523.1 | - | 15 - 16 | 1805 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |