Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792736 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792774 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792738 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792771 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792735 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792739 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792772 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536293 |
AK140117.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536293 |
BC052068.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536293 |
NM_001004147.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
533264 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
277937 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
95419 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
179690 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
16532 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140977 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792745 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792740 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792731 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792732 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792748 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792755 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792754 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792727 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792709 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792744 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792739 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792750 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792726 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792729 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792747 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792711 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792723 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792707 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792738 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792742 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792706 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792716 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792730 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792756 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792718 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792736 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792722 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792721 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792704 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792715 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30792734 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08613169 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P140977 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08613154 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P257258 |
D16333.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P135878 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P140977 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12621191 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12621199 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12621188 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12621192 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12621194 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792772 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792771 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792739 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792736 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792738 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792735 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30792774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30792841 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12621192 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792754 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792711 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792752 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792715 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792718 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792701 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792706 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12604174 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792745 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792795 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792695 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792768 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792762 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792734 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792721 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792790 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792730 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792784 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792709 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12942307 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12787247 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792755 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792742 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12741499 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792787 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12621206 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792783 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792764 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792693 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792772 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792727 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792700 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792794 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792771 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792697 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792786 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792796 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792747 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792797 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792785 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12621188 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792739 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792740 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792791 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792758 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792781 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792779 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792793 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792707 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792759 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792722 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792750 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792789 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792770 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13003092 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30792774 |
- |
- |