Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007824.2 | NP_031850.2 | 3 - 4 | 968 | 63 | |
GenBank mRNA | AK050020.1 | - | 3 - 4 | 968 | 63 | |
AK050210.1 | - | 3 - 4 | 969 | 63 | ||
AK050220.1 | - | 3 - 4 | 968 | 63 | ||
AK050260.1 | - | 3 - 4 | 968 | 63 | ||
BC130261.1 | - | 3 - 4 | 938 | 40142789 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |