Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008564.2 | NP_032590.2 | 9 - 10 | 1650 | 62 | |
GenBank mRNA | AF004105.1 | - | 8 - 9 | 1540 | 62 | |
AK037751.1 | - | 9 - 10 | 1575 | 62 | ||
AK077801.1 | - | 9 - 10 | 1557 | 62 | ||
AK088156.1 | - | 9 - 10 | 1563 | 62 | ||
AK088378.1 | - | 9 - 10 | 1550 | 62 | ||
AK129039.1 | - | 9 - 10 | 1531 | 62 | ||
AK145176.1 | - | 9 - 10 | 1562 | 62 | ||
AK146187.1 | - | 9 - 10 | 1565 | 62 | ||
AK146375.1 | - | 9 - 10 | 1574 | 62 | ||
AK191042.1 | - | 2 - 3 | 141 | 62 | ||
BC055318.1 | - | 9 - 10 | 1559 | 5696348 | 62 | |
D86725.1 | - | 9 - 10 | 1570 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |