Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_001093775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_001093775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_001093776.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_001093776.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_001093778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_001093778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_008666.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
NM_008666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001874.mm.1 |
TC1200000247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_001093775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_001093775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_001093776.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_001093776.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_001093778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_001093778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_008666.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
NM_008666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001876.mm.1 |
TC1200000247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001873.mm.1 |
NM_001093776.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001873.mm.1 |
NM_001093776.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001873.mm.1 |
TC1200000247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_001093775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_001093775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_001093776.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_001093776.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_001093778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_001093778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_008666.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
NM_008666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001875.mm.1 |
TC1200000247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001870.mm.1 |
NM_001093775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001870.mm.1 |
NM_001093775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001870.mm.1 |
NM_001093778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001870.mm.1 |
NM_001093778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001870.mm.1 |
NM_008666.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001870.mm.1 |
NM_008666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001870.mm.1 |
TC1200000247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_001093775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_001093775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_001093776.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_001093776.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_001093778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_001093778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_008666.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
NM_008666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001878.mm.1 |
TC1200000247.mm.1 |
MTA TC |