Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad54l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122958.1 | NP_001116430.1 | 16 - 17 | 1989 | 98 | |
NM_001122959.1 | NP_001116431.1 | 16 - 17 | 1968 | 98 | ||
NM_009015.3 | NP_033041.3 | 15 - 16 | 2319 | 98 | ||
GenBank mRNA | AK030753.1 | - | 16 - 17 | 1947 | 98 | |
AK088876.1 | - | 16 - 17 | 1986 | 98 | ||
AK161644.1 | - | 16 - 17 | 1942 | 98 | ||
BC021643.1 | - | 15 - 16 | 2300 | 3987790 | 98 | |
X97796.1 | - | 15 - 16 | 1779 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |