Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009043.1 | NP_033069.1 | 2 - 3 | 137 | 95 | |
GenBank mRNA | AK008196.1 | - | 2 - 3 | 136 | 95 | |
BC098503.1 | - | 1 - 2 | 120 | 6431377 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |