Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037761.2 | NP_001032850.1 | 4 - 5 | 458 | 60 | |
NM_001271405.1 | NP_001258334.1 | 4 - 5 | 437 | 60 | ||
NM_001271406.1 | NP_001258335.1 | 4 - 5 | 423 | 60 | ||
NM_009798.4 | NP_033928.1 | 4 - 5 | 458 | 60 | ||
GenBank mRNA | AK156778.1 | - | 4 - 5 | 427 | 60 | |
AK159390.1 | - | 4 - 5 | 432 | 60 | ||
AK160082.1 | - | 4 - 5 | 433 | 60 | ||
AK162757.1 | - | 4 - 5 | 425 | 60 | ||
AK168738.1 | - | 4 - 5 | 386 | 60 | ||
BC002053.1 | - | 4 - 5 | 400 | 3590980 | 60 | |
FJ692320.1 | - | 4 - 5 | 423 | 60 | ||
U10406.1 | - | 4 - 5 | 378 | 60 | ||
U10407.1 | - | 4 - 5 | 349 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |