Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161623.1 | NP_001155095.1 | 7 - 8 | 586 | 61 | |
NM_009862.2 | NP_033992.2 | 9 - 10 | 963 | 61 | ||
GenBank mRNA | AF081536.1 | - | 9 - 10 | 963 | 61 | |
AF081537.1 | - | 8 - 9 | 662 | 61 | ||
AF081538.1 | - | 8 - 9 | 662 | 61 | ||
AF098068.1 | - | 8 - 9 | 709 | 61 | ||
AJ223729.1 | - | 8 - 9 | 683 | 61 | ||
AK147013.1 | - | 8 - 9 | 680 | 61 | ||
AK167850.1 | - | 7 - 8 | 574 | 61 | ||
BC028635.1 | - | 8 - 9 | 716 | 1262927 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |