Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 7 - 8 | 1448 | 91 | |
NM_001110351.1 | NP_001103821.1 | 8 - 9 | 1325 | 91 | ||
NM_011378.2 | NP_035508.2 | 8 - 9 | 1556 | 91 | ||
GenBank mRNA | AK032209.1 | - | 7 - 8 | 1316 | 91 | |
AK136347.1 | - | 2 - 3 | 250 | 91 | ||
AK220292.1 | - | 7 - 8 | 1425 | 91 | ||
BC052716.1 | - | 7 - 8 | 1335 | 6837144 | 91 | |
BC053385.1 | - | 7 - 8 | 1318 | 91 | ||
BC078454.1 | - | 7 - 8 | 1309 | 91 | ||
L36831.1 | - | 7 - 8 | 1239 | 91 | ||
L38620.2 | - | 6 - 7 | 1215 | 91 | ||
L38621.2 | - | 6 - 7 | 1215 | 91 | ||
U22394.1 | - | 8 - 9 | 1564 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |