Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077591.1 | NP_001071059.1 | 3 - 4 | 532 | 63 | |
NM_011651.2 | NP_035781.2 | 3 - 4 | 532 | 63 | ||
GenBank mRNA | AF025310.2 | - | 3 - 4 | 512 | 63 | |
BC118618.1 | - | 3 - 4 | 511 | 40107271 | 63 | |
BC121827.1 | - | 3 - 4 | 511 | 40107270 | 63 | |
CN831975.1 | - | 255 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |