Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011992.2 | NP_036122.2 | 5 - 6 | 876 | 134 | |
GenBank mRNA | AF049125.1 | - | 5 - 6 | 865 | 134 | |
AK041996.1 | - | 873 | 134 | |||
AK077486.1 | - | 5 - 6 | 852 | 134 | ||
AK169765.1 | - | 5 - 6 | 850 | 134 | ||
BC132320.1 | - | 5 - 6 | 841 | 40130597 | 134 | |
BC145668.1 | - | 5 - 6 | 841 | 40130971 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |