Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016782.2 | NP_058062.2 | 8 - 9 | 1453 | 57 | |
GenBank mRNA | AF039833.1 | - | 8 - 9 | 1463 | 57 | |
AK154468.1 | - | 2214 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |