Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166531.1 | NP_001160003.1 | 3 - 4 | 304 | 111 | |
NM_001166532.1 | NP_001160004.1 | 2 - 3 | 398 | 111 | ||
NM_019460.2 | NP_062333.1 | 2 - 3 | 255 | 111 | ||
GenBank mRNA | AB032165.1 | - | 1 - 2 | 100 | 111 | |
AK142330.1 | - | 2 - 3 | 283 | 111 | ||
AK154826.1 | - | 2 - 3 | 400 | 111 | ||
AK160573.1 | - | 3 - 4 | 304 | 111 | ||
BC018217.1 | - | 2 - 3 | 266 | 111 | ||
BC036972.1 | - | 2 - 3 | 325 | 111 | ||
BC066188.1 | - | 2 - 3 | 208 | 30476111 | 111 | |
CJ077633.1 | - | 2 - 3 | 255 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |