Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111058.1 | NP_001104528.1 | 1 - 2 | 114 | 64 | |
NM_021293.3 | NP_067268.1 | 1 - 2 | 114 | 64 | ||
GenBank mRNA | AK046303.1 | - | 3 - 4 | 337 | 64 | |
AK089205.1 | - | 1 - 2 | 31 | 64 | ||
AK155745.1 | - | 1 - 2 | 43 | 64 | ||
AK170937.1 | - | 1 - 2 | 82 | 64 | ||
BC132379.1 | - | 1 - 2 | 114 | 40130656 | 64 | |
S71345.1 | - | 1 - 2 | 45 | 64 | ||
S71403.1 | - | 1 - 2 | 45 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |