Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271472.1 | NP_001258401.1 | 22 - 23 | 3036 | 77 | |
NM_001271473.1 | NP_001258402.1 | 19 - 20 | 2703 | 77 | ||
NM_022723.3 | NP_073560.2 | 21 - 22 | 2946 | 77 | ||
GenBank mRNA | AF276425.1 | - | 20 - 21 | 3239 | 77 | |
AK041590.1 | - | 21 - 22 | 2941 | 77 | ||
AK083722.1 | - | 5 - 6 | 895 | 77 | ||
BC066066.1 | - | 22 - 23 | 2994 | 6850999 | 77 | |
BC080278.1 | - | 19 - 20 | 2671 | 30354974 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |