Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 26 - 27 | 3483 | 117 | |
GenBank mRNA | AK135430.1 | - | 26 - 27 | 2977 | 117 | |
AK136346.1 | - | 7 - 8 | 724 | 117 | ||
BC050946.1 | - | 26 - 27 | 3421 | 6822283 | 117 | |
BC054537.1 | - | 26 - 27 | 3405 | 117 | ||
D12646.1 | - | 26 - 27 | 3810 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |