Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB011550.1 | - | 10 - 11 | 1159 | 96 | |
AK076365.1 | - | 8 - 9 | 2075 | 96 | ||
AK133474.1 | - | 10 - 11 | 1127 | 96 | ||
AK139088.1 | - | 10 - 11 | 1365 | 96 | ||
AK144793.1 | - | 10 - 11 | 1343 | 96 | ||
AK155268.1 | - | 7 - 8 | 2204 | 96 | ||
BC015077.1 | - | 10 - 11 | 1041 | 96 | ||
BC145595.1 | - | 9 - 10 | 1133 | 96 | ||
BC145596.1 | - | 10 - 11 | 1048 | 96 | ||
U81490.1 | - | 10 - 11 | 1099 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |