Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00003616.mm.1 |
NM_009575.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003616.mm.1 |
NM_009575.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003616.mm.1 |
TC0X00000538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003619.mm.1 |
NM_009575.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003619.mm.1 |
NM_009575.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003619.mm.1 |
TC0X00000538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003613.mm.1 |
NM_009575.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003613.mm.1 |
NM_009575.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003613.mm.1 |
TC0X00000538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003618.mm.1 |
NM_009575.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003618.mm.1 |
NM_009575.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003618.mm.1 |
TC0X00000538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003617.mm.1 |
NM_009575.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003617.mm.1 |
NM_009575.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003617.mm.1 |
TC0X00000538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003620.mm.1 |
NM_009575.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003620.mm.1 |
NM_009575.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003620.mm.1 |
TC0X00000538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003621.mm.1 |
NM_009575.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003621.mm.1 |
NM_009575.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003621.mm.1 |
TC0X00000538.mm.1 |
MTA TC |