Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021901.3 | NP_068701.1 | 1 - 2 | 778 | 61 | |
GenBank mRNA | BC018246.1 | - | 1 - 2 | 778 | 4164519 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |