Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024188.6 | NP_077150.1 | 11 - 12 | 1447 | 62 | |
GenBank mRNA | AB085609.1 | - | 11 - 12 | 1189 | 62 | |
AK009027.1 | - | 11 - 12 | 1227 | 62 | ||
AK011354.1 | - | 11 - 12 | 1127 | 62 | ||
AK078086.1 | - | 11 - 12 | 1445 | 62 | ||
AK146158.1 | - | 11 - 12 | 1205 | 62 | ||
AK146342.1 | - | 11 - 12 | 1192 | 62 | ||
AK151662.1 | - | 11 - 12 | 1186 | 62 | ||
AK151695.1 | - | 11 - 12 | 1177 | 62 | ||
AK167681.1 | - | 11 - 12 | 1277 | 62 | ||
AK215681.1 | - | 4 - 5 | 303 | 62 | ||
BC003422.1 | - | 11 - 12 | 1151 | 2598824 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |