Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colec12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130449.2 | NP_569716.2 | 4 - 5 | 384 | 90 | |
GenBank mRNA | AB038519.1 | - | 4 - 5 | 355 | 90 | |
AB078434.1 | - | 4 - 5 | 370 | 90 | ||
AK042772.1 | - | 4 - 5 | 385 | 90 | ||
AK141928.1 | - | 4 - 5 | 370 | 90 | ||
AK158366.1 | - | 4 - 5 | 406 | 90 | ||
BC009162.1 | - | 4 - 5 | 373 | 90 | ||
BC057936.1 | - | 4 - 5 | 373 | 5326386 | 90 | |
BC060657.1 | - | 4 - 5 | 363 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |