Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008485.3 | NP_032511.3 | 10 - 11 | 1599 | 99 | |
GenBank mRNA | U43327.2 | - | 9 - 10 | 1332 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |