Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025310.2 | NP_079586.1 | 11 - 12 | 1059 | 99 | |
GenBank mRNA | AK004996.2 | - | 11 - 12 | 1059 | 99 | |
AK031514.1 | - | 11 - 12 | 1046 | 99 | ||
AK145490.1 | - | 11 - 12 | 1084 | 99 | ||
BC012281.1 | - | 11 - 12 | 1056 | 4012961 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |