Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286560.1 | NP_001273489.1 | 7 - 8 | 898 | 91 | |
NM_026168.4 | NP_080444.1 | 8 - 9 | 1007 | 91 | ||
NM_026355.3 | NP_080631.1 | 8 - 9 | 1007 | 91 | ||
GenBank mRNA | AK004661.1 | - | 8 - 9 | 671 | 91 | |
AK007498.1 | - | 8 - 9 | 656 | 91 | ||
AK008971.1 | - | 8 - 9 | 663 | 91 | ||
AK009483.1 | - | 8 - 9 | 659 | 91 | ||
AK010529.1 | - | 8 - 9 | 656 | 91 | ||
AK016275.1 | - | 8 - 9 | 1008 | 91 | ||
AK017876.1 | - | 8 - 9 | 691 | 91 | ||
AK030045.1 | - | 8 - 9 | 657 | 91 | ||
AK086727.1 | - | 8 - 9 | 659 | 91 | ||
AK135968.1 | - | 5 - 6 | 344 | 91 | ||
AK162267.1 | - | 7 - 8 | 631 | 91 | ||
AK167391.1 | - | 8 - 9 | 660 | 91 | ||
BC006895.1 | - | 8 - 9 | 625 | 2651322 | 91 | |
BC018188.1 | - | 8 - 9 | 659 | 3711066 | 91 | |
BC026558.1 | - | 8 - 9 | 619 | 5030661 | 91 | |
BC064749.1 | - | 8 - 9 | 720 | 30046220 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |