Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028301.4 | NP_082577.2 | 10 - 11 | 1404 | 64 | |
GenBank mRNA | AK012494.1 | - | 10 - 11 | 1353 | 64 | |
AK089037.1 | - | 10 - 11 | 1379 | 64 | ||
AK177037.1 | - | 2 - 3 | 285 | 64 | ||
BC022691.1 | - | 2 - 3 | 260 | 64 | ||
BC039644.1 | - | 3 - 4 | 357 | 64 | ||
BC050929.1 | - | 10 - 11 | 1340 | 6807194 | 64 | |
BC051143.1 | - | 10 - 11 | 1371 | 6531161 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |