Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031376.3 | NP_113553.1 | 2 - 3 | 538 | 57 | |
GenBank mRNA | AF293806.1 | - | 2 - 3 | 538 | 57 | |
AK089785.1 | - | 2 - 3 | 571 | 57 | ||
AK154326.1 | - | 2 - 3 | 566 | 57 | ||
AK170130.1 | - | 2 - 3 | 571 | 57 | ||
AK170925.1 | - | 2 - 3 | 571 | 57 | ||
AK171021.1 | - | 2 - 3 | 566 | 57 | ||
BC113141.1 | - | 2 - 3 | 497 | 40047744 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |