Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133770.2 | NP_598531.2 | 13 - 14 | 1382 | 68 | |
GenBank mRNA | AK002619.1 | - | 13 - 14 | 1372 | 68 | |
AK079366.1 | - | 3 - 4 | 426 | 68 | ||
BC013485.1 | - | 10 - 11 | 945 | 4242474 | 68 | |
BC083324.1 | - | 11 - 12 | 1183 | 6404832 | 68 | |
BC093498.1 | - | 12 - 13 | 1319 | 3669346 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |