Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200005123.mm.1 |
NM_001080968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005123.mm.1 |
NM_001080968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005123.mm.1 |
NM_133852.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005123.mm.1 |
NM_133852.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005123.mm.1 |
TC0200000574.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005119.mm.1 |
NM_001080968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005119.mm.1 |
NM_001080968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005119.mm.1 |
NM_133852.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005119.mm.1 |
NM_133852.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005119.mm.1 |
TC0200000574.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005120.mm.1 |
NM_001080968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005120.mm.1 |
NM_001080968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005120.mm.1 |
NM_133852.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005120.mm.1 |
NM_133852.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005120.mm.1 |
TC0200000574.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005122.mm.1 |
NM_001080968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005122.mm.1 |
NM_001080968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005122.mm.1 |
NM_133852.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005122.mm.1 |
NM_133852.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005122.mm.1 |
TC0200000574.mm.1 |
MTA TC |