Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133925.2 | NP_598686.2 | 12 - 13 | 1421 | 65 | |
GenBank mRNA | AK156773.1 | - | 12 - 13 | 1358 | 65 | |
AK161670.1 | - | 12 - 13 | 1388 | 65 | ||
AK167050.1 | - | 12 - 13 | 1423 | 65 | ||
BC018373.1 | - | 8 - 9 | 795 | 3708613 | 65 | |
BC023293.1 | - | 2 - 3 | 170 | 65 | ||
BC024602.1 | - | 12 - 13 | 1302 | 65 | ||
BC027123.1 | - | 5 - 6 | 727 | 5356457 | 65 | |
BC040811.1 | - | 12 - 13 | 1366 | 4015830 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |