Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028078.3 | NP_082354.1 | 3 - 4 | 478 | 60 | |
GenBank mRNA | AK008060.1 | - | 3 - 4 | 449 | 60 | |
AK078934.1 | - | 3 - 4 | 449 | 60 | ||
BC004806.1 | - | 3 - 4 | 431 | 3584645 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |