Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028787.4 | NP_083063.2 | 2 - 3 | 358 | 88 | |
GenBank mRNA | AK004892.2 | - | 2 - 3 | 339 | 88 | |
AK145848.1 | - | 2 - 3 | 359 | 88 | ||
AK169012.1 | - | 2 - 3 | 220 | 88 | ||
AK171951.1 | - | 2 - 3 | 332 | 88 | ||
BC026858.1 | - | 2 - 3 | 217 | 4014064 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |