Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010442.2 | NP_034572.1 | 2 - 3 | 279 | 69 | |
GenBank mRNA | AK150934.1 | - | 2 - 3 | 276 | 69 | |
AK151005.1 | - | 2 - 3 | 276 | 69 | ||
AK151627.1 | - | 2 - 3 | 254 | 69 | ||
AK153420.1 | - | 2 - 3 | 276 | 69 | ||
AK153516.1 | - | 2 - 3 | 276 | 69 | ||
AK153566.1 | - | 2 - 3 | 276 | 69 | ||
AK154598.1 | - | 2 - 3 | 277 | 69 | ||
AK159521.1 | - | 2 - 3 | 276 | 69 | ||
AK159870.1 | - | 2 - 3 | 280 | 69 | ||
AK159959.1 | - | 2 - 3 | 280 | 69 | ||
AK169227.1 | - | 2 - 3 | 276 | 69 | ||
BC010757.1 | - | 2 - 3 | 248 | 4235374 | 69 | |
BY319630.1 | - | 2 - 3 | 281 | 69 | ||
M33203.1 | - | 2 - 3 | 221 | 69 | ||
X13356.1 | - | 2 - 3 | 221 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |