Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011801.1 | NP_035931.1 | 3 - 4 | 493 | 78 | |
GenBank mRNA | AB010828.1 | - | 3 - 4 | 493 | 78 | |
AK133989.1 | - | 3 - 4 | 511 | 78 | ||
AK160532.1 | - | 3 - 4 | 458 | 78 | ||
BC005589.1 | - | 3 - 4 | 474 | 3964401 | 78 | |
Y08219.1 | - | 3 - 4 | 492 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |