Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018827.2 | NP_061297.1 | 7 - 8 | 1346 | 57 | |
GenBank mRNA | AB040038.1 | - | 7 - 8 | 1346 | 57 | |
AK019030.1 | - | 2 - 3 | 339 | 57 | ||
AK160445.1 | - | 2 - 3 | 305 | 57 | ||
BC107376.1 | - | 2 - 3 | 175 | 57 | ||
BC107377.1 | - | 2 - 3 | 200 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |