Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025816.3 | NP_080092.2 | 7 - 8 | 977 | 92 | |
GenBank mRNA | AK004574.1 | - | 7 - 8 | 959 | 92 | |
AK146076.1 | - | 7 - 8 | 957 | 92 | ||
BC014798.1 | - | 7 - 8 | 898 | 3962810 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |