Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2077353 |
BC099443.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2077353 |
NM_001162524.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041624 |
NM_026087.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041624 |
AK005400.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041624 |
AK145926.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041626 |
BC099443.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041626 |
NM_026087.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041626 |
AK005400.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041626 |
AK005565.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041626 |
AK131656.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
AK007156.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
AK015996.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
AK016294.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
AK029665.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
AY243470.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
BC116805.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
BC116809.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793035 |
NM_026458.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
261142 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
712629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
103313 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
706835 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
83493 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P176432 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041624 |
NM_026087.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041624 |
AK005400.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041624 |
AK145926.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041626 |
BC099443.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041626 |
NM_026087.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041626 |
AK005400.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041626 |
AK005565.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041626 |
AK131656.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2077353 |
BC099443.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2077353 |
NM_001162524.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987457 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987402 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987458 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987423 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987436 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987413 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987411 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987429 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987426 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987420 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987456 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987451 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987440 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987443 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987422 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987406 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987433 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987434 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987447 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987396 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987427 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987438 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987397 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987454 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987409 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987445 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987403 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987416 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987408 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24987418 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P356505 |
BC099443.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356505 |
NM_026087.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356505 |
AK005400.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356505 |
AK005565.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356505 |
AK131656.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P100488 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P110904 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05916061 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05916058 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06173274 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06240879 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06240875 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06240890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987393 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987385 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24987447 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987427 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05916061 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987409 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987388 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987426 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987420 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987398 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06240879 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987497 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987493 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987476 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987484 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987445 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06147598 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987482 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987393 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06661206 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987411 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987423 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987396 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06240875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987487 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987397 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987406 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06222587 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987443 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05916058 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987408 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987468 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987492 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06654065 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987456 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987479 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987402 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06654056 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06222590 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987447 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06240888 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06222611 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06240890 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987403 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06254161 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987467 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06173274 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987460 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987441 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06240877 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987499 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987495 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987457 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987464 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987491 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06222614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987455 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987416 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06240894 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05916063 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24987390 |
- |
- |