Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029943.1 | NP_084219.1 | 3 - 4 | 490 | 68 | |
GenBank mRNA | AB072498.1 | - | 3 - 4 | 490 | 68 | |
AK021248.1 | - | 3 - 4 | 472 | 68 | ||
AK033489.1 | - | 3 - 4 | 482 | 68 | ||
AK040145.1 | - | 3 - 4 | 424 | 68 | ||
AK050858.1 | - | 3 - 4 | 453 | 68 | ||
AK080916.1 | - | 702 | 68 | |||
AK081677.1 | - | 3 - 4 | 445 | 68 | ||
AK088918.1 | - | 3 - 4 | 459 | 68 | ||
BC026769.1 | - | 3 - 4 | 422 | 68 | ||
BC053107.1 | - | 2 - 3 | 1222 | 68 | ||
BC078633.1 | - | 3 - 4 | 448 | 6488060 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |