Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK050014.1 | - | 10 - 11 | 1083 | 56 | |
AK075771.1 | - | 10 - 11 | 1070 | 56 | ||
AK075791.1 | - | 10 - 11 | 1069 | 56 | ||
AK077853.1 | - | 10 - 11 | 1001 | 56 | ||
AK149765.1 | - | 10 - 11 | 1097 | 56 | ||
AK153951.1 | - | 10 - 11 | 988 | 56 | ||
AK161834.1 | - | 8 - 9 | 928 | 56 | ||
AK165494.1 | - | 9 - 10 | 1020 | 56 | ||
AK166476.1 | - | 10 - 11 | 1097 | 56 | ||
AK187835.1 | - | 2 - 3 | 177 | 56 | ||
BC002318.1 | - | 10 - 11 | 1067 | 3593792 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |