Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030566.2 | NP_085043.2 | 3 - 4 | 757 | 69 | |
GenBank mRNA | AK089938.1 | - | 3 - 4 | 461 | 69 | |
AK144836.1 | - | 3 - 4 | 443 | 69 | ||
AK157832.1 | - | 3 - 4 | 418 | 69 | ||
BC004088.1 | - | 2 - 3 | 503 | 3593722 | 69 | |
BC015287.1 | - | 3 - 4 | 765 | 4240442 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |